
Scutellarin

Synonym(s):
4′,5,6,7-Tetrahydroxyflavone 7-glucuronide, 5,6-Dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-1-benzopyran-7-yl-β-D-glucopyranosiduronic acid, 7-(β-D-Glucopyranuronosyloxy)-5,6-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Breviscapin, Scutellarein-7-O-β-D-glucuronide
Empirical Formula (Hill Notation):
C21H18O12
CAS Number:
Molecular Weight:
462.36
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing