Skip to Content
Merck
(S)-(−)-1,2,4-Butanetriol 98%

(S)-(−)-1,2,4-Butanetriol

(S)-(−)-1,2,4-Butanetriol 98%
Linear Formula:
HOCH2CH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
106.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing