
Rhodium(II) octanoate, dimer

Linear Formula:
[[CH3(CH2)6CO2]2Rh]2
CAS Number:
Molecular Weight:
778.63
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing