
Resorufin pentyl ether

Synonym(s):
7-Pentyloxy-3-phenoxazone, 7-Pentyloxy-3H-phenoxazin-3-one, O7-Pentylresorufin, Pentoxyresorufin
Empirical Formula (Hill Notation):
C17H17NO3
CAS Number:
Molecular Weight:
283.32
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1