Skip to Content
Merck
Repaglinide European Pharmacopoeia (EP) Reference Standard

Repaglinide

Repaglinide European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(S)-(+)-2-Ethoxy-4-[N-[1-(2-piperidinophenyl)-3-methyl-1-butyl]aminocarbonylmethyl]benzoic acid, Novonorm, Prandin, Repaglinide
Empirical Formula (Hill Notation):
C27H36N2O4
CAS Number:
Molecular Weight:
452.59

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4