Skip to Content
Merck
(R)-(−)-3-Quinuclidinol hydrochloride 97%

(R)-(−)-3-Quinuclidinol hydrochloride

(R)-(−)-3-Quinuclidinol hydrochloride 97%
Synonym(s):
(R)-(−)-3-Hydroxyquinuclidine hydrochloride
Empirical Formula (Hill Notation):
C7H13NO · HCl
CAS Number:
Molecular Weight:
163.65

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing