Skip to Content
BM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
aseptic filling bell
Synonym(s):
Corning
®
3965
Products
Product Category
bottles (4)
Manufacturer
OD (mm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS396570
Pyrex
®
aseptic filling bell,
hose outlet O.D. 11.0 mm, O.D. × H 73 mm × 130 mm
Expand
View Pricing
CLS396520
Pyrex
®
aseptic filling bell,
hose outlet O.D. 7.0 mm
Expand
View Pricing
CLS396550
Pyrex
®
aseptic filling bell,
hose outlet O.D. 11.0 mm, O.D. × H 51 mm × 95 mm
Expand
View Pricing
CLS3965100
Pyrex
®
aseptic filling bell,
hose outlet O.D. 12.7 mm
Expand
View Pricing