Pseudomonas paraeruginosa WDCM 00026 Vitroids™
Synonym(s):
Pseudomonas aeruginosa WDCM 00026 Vitroids™
Products
Brand
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing