
Prothioconazole

Synonym(s):
2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2-hydroxypropyl]-1,2-dihydro-3H-1,2,4-triazole-3-thione
Empirical Formula (Hill Notation):
C14H15Cl2N3OS
CAS Number:
Molecular Weight:
344.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2