Skip to Content
Merck
Prazepam European Pharmacopoeia (EP) Reference Standard

Prazepam

Prazepam European Pharmacopoeia (EP) Reference Standard
Synonym(s):
7-Chloro-1-(cyclopropylmethyl)-1,3-dihydro-5-phenyl-2H-1,4-benzodiazepin-2-one
Empirical Formula (Hill Notation):
C19H17ClN2O
CAS Number:
Molecular Weight:
324.80

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3