
Potassium trichloroammineplatinate

Synonym(s):
Potassium aminetrichloroplatinate(II), Magnus red anion salt, Potassium trichloroammineplatinate(II)
Linear Formula:
KPt(NH3)Cl3
CAS Number:
Molecular Weight:
357.57
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2