Skip to Content
BM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium permanganate solution
Empirical Formula (Hill Notation):
KMnO
4
CAS Number:
7722-64-7
Molecular Weight:
158.03
Products
Brand
Sigma-Aldrich (6)
Formula Weight
Physical Form
Reagent Type
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
24-5170
Potassium permanganate solution,
0.02 M
Expand
View Pricing
24-5180
Potassium permanganate solution,
5 mM
Expand
View Pricing
24-5200
Potassium permanganate solution,
2 mM
Expand
View Pricing
24-5165
Potassium permanganate solution,
0.1 M
Expand
View Pricing
24-5160
Potassium permanganate solution,
0.2 M
Expand
View Pricing
24-5175
Potassium permanganate solution,
0.01 M
Expand
View Pricing