![Pimaricin preparation ~2.5% (γ-irradiated Pimaricin), aqueous suspension](/deepweb/assets/sigmaaldrich/product/structures/344/194/76321063-4059-4807-b11c-970ed369927f/640/76321063-4059-4807-b11c-970ed369927f.png)
Pimaricin preparation
![Pimaricin preparation ~2.5% (γ-irradiated Pimaricin), aqueous suspension](/deepweb/assets/sigmaaldrich/product/structures/344/194/76321063-4059-4807-b11c-970ed369927f/640/76321063-4059-4807-b11c-970ed369927f.png)
Synonym(s):
Natamycin preparation
Empirical Formula (Hill Notation):
C33H47NO13
CAS Number:
Molecular Weight:
665.73
Products
Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing