
Phthalic acid

Synonym(s):
Phthalic acid, 1,2-Benzenedicarboxylic acid
Linear Formula:
C6H4-1,2-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing