Skip to Content
BM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Perflubron
Synonym(s):
1-Bromoheptadecafluorooctane, Heptadecafluorooctyl bromide, Perfluorooctyl bromide
Linear Formula:
CF
3
(CF
2
)
7
Br
CAS Number:
423-55-2
Molecular Weight:
498.96
Products
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1510801
Perflubron,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
PHR3219
Perflubron,
pharmaceutical secondary standard, certified reference material
Expand
View Pricing