
Orcinol β-D-glucoside

Synonym(s):
Sakakin, 1-O-β-D-Glucosylorcinol
Empirical Formula (Hill Notation):
C13H18O7
CAS Number:
Molecular Weight:
286.28
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing