
Oleyl oleate

Synonym(s):
Oleic acid oleyl ester
Empirical Formula (Hill Notation):
C36H68O2
CAS Number:
Molecular Weight:
532.92
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Functional Group
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing