Octyl gallate
Synonym(s):
Octyl gallate
Linear Formula:
3,4,5-(HO)3C6H2CO2(CH2)7CH3
CAS Number:
Molecular Weight:
282.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3