Skip to Content
Merck
Octyl β-D-glucopyranoside BioXtra, ≥98% (GC)

Octyl β-D-glucopyranoside

Octyl β-D-glucopyranoside BioXtra, ≥98% (GC)
Synonym(s):
n-Octyl-β-D-glucopyranoside, ULTROL® Grade, n-Octyl glucoside, OGP
Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37
EC Number:

Products

Product Category
Brand
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment