Skip to Content
Merck
N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt plasmin substrate

N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt

N-(p-Tosyl)-Gly-Pro-Lys 4-nitroanilide acetate salt plasmin substrate
Synonym(s):
N-Tosylglycyl-L-prolyl-L-lysine 4-nitroanilide acetate salt
Linear Formula:
C26H34N6O7S · C2H4O2
CAS Number:
Molecular Weight:
634.70
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment