Skip to Content
Merck
Nickel(II) chloride hexahydrate ReagentPlus®

Nickel(II) chloride hexahydrate

Nickel(II) chloride hexahydrate ReagentPlus®
Synonym(s):
Nickel(II) chloride hexahydrate
Linear Formula:
NiCl2 · 6H2O
CAS Number:
Molecular Weight:
237.69

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Technique