
N-Formyl-L-leucine (3S,4R,6S)-3-hexyl-2-oxo-6-undecyltetrahydro-2H-pyran-4-yl ester

Empirical Formula (Hill Notation):
C29H53NO5
CAS Number:
Molecular Weight:
495.73
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing