
Muramic acid

Synonym(s):
2-Amino-3-O-(1-carboxyethyl)-2-deoxy-D-glucose
Empirical Formula (Hill Notation):
C9H17NO7
CAS Number:
Molecular Weight:
251.23
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing