
Morin

Synonym(s):
2′,3,4′,5,7-Pentahydroxyflavone
Empirical Formula (Hill Notation):
C15H10O7
CAS Number:
Molecular Weight:
302.24
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing