Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing