
Mometasone furoate for system suitability

Synonym(s):
Mometasone furoate, (11β,16α)-9,21-Dichloro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C27H30Cl2O6
CAS Number:
Molecular Weight:
521.43
Products
Formula Weight
Gene Alias
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing