
Metformin Related Compound C

Synonym(s):
N,N-Dimethyl-1,3,5-triazine-2,4,6-triamine
Empirical Formula (Hill Notation):
C5H10N6
CAS Number:
Molecular Weight:
154.17
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2