
Meranzin hydrate

Synonym(s):
8-[(2S)-2,3-Dihydroxy-3-methylbutyl]-7-methoxy-2H-1-benzopyran-2-one
Empirical Formula (Hill Notation):
C15H18O5
CAS Number:
Molecular Weight:
278.30
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Reference Material Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing