Skip to Content
Merck
Lucifer Yellow CH dilithium salt fluorescent stain

Lucifer Yellow CH dilithium salt

Lucifer Yellow CH dilithium salt fluorescent stain
Synonym(s):
6-Amino-2,3-dihydro-1,3-dioxo-2-hydrazinocarbonylamino-1H-benz[d,e]isoquinoline-5,8-disulfonic acid dilithium salt
Empirical Formula (Hill Notation):
C13H9Li2N5O9S2
CAS Number:
Molecular Weight:
457.25
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing