
L-Histidine monohydrochloride monohydrate

Synonym(s):
L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride
Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Optical Isomer
Amino Acid
Reference Material Type
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing