Skip to Content
Merck
L-(+)-Tartaric acid SAJ first grade, ≥99.5%

L(+)-Tartaric acid

L-(+)-Tartaric acid SAJ first grade, ≥99.5%
Synonym(s):
L(+)-Tartaric acid, (2R,3R)-(+)-Tartaric acid, L-Threaric acid, L-(+)-Tartaric acid
Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Special Grade
Technique