Skip to Content
Merck
Isoflucypram PESTANAL®, analytical standard

Isoflucypram

Isoflucypram PESTANAL®, analytical standard
Synonym(s):
N-[[5-Chloro-2-(1-methylethyl)phenyl]methyl]-N-cyclopropyl-3-(difluoromethyl)-5-fluoro-1-methyl-1H-pyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C19H21ClF3N3O
CAS Number:
Molecular Weight:
399.84

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing