
Irinotecan hydrochloride

Synonym(s):
Irinotecan monohydrochloride trihydrate, [1,4′-Bipiperidine]-1′-carboxylic acid (S)-4,11-diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3′,4′:6,7]indolizino[1,2-b]quinolin-9-yl ester monohydrochloride trihydrate
Empirical Formula (Hill Notation):
C33H38N4O6 · HCl · 3H2O
CAS Number:
Molecular Weight:
677.18
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2