
Irbesartan impurity A

Synonym(s):
1-[(1-Oxopentyl)amino]-N-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]cyclopentanecarboxamide
Empirical Formula (Hill Notation):
C25H30N6O2
CAS Number:
Molecular Weight:
446.54
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing