Skip to Content
Merck
Imino-tris(dimethylamino)phosphorane 97%

Imino-tris(dimethylamino)phosphorane

Imino-tris(dimethylamino)phosphorane 97%
Synonym(s):
N,N,N′,N′,N″,N″-Hexamethylphosphorimidic triamide, Phosphazene base P1-H
Linear Formula:
[(CH3)2N]3P=NH
CAS Number:
Molecular Weight:
178.22

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing