
Hydroxyphenyl fluorescein solution

Synonym(s):
Hydroxyphenyl fluorescein, 2-[6-(4′-hydroxy)phenoxy-3H-xanthen-3-on-9-yl]benzoic acid, HPF
Empirical Formula (Hill Notation):
C26H16O6
Molecular Weight:
424.40
EC Number:
Products
Product Category
Brand
Formula Weight
Showing 1-1 of 1
Compare
Product Number
Description
Pricing