Skip to Content
Merck
Hydrogen peroxide solution contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis

Hydrogen peroxide solution

Hydrogen peroxide solution contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Synonym(s):
Hydrogen peroxide solution
Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Purity
Agency
Research Area
Quality Segment
Application
Markush Class
Markush Group
Microorganism Suitability
Reagent Type
Special Grade
Technique
Showing 1-19 of 19
Compare
Product Number
Description
Pricing