
Hydrocodone bitartrate

Synonym(s):
Hydrocodone bitartrate salt hemi(pentahydrate)
Empirical Formula (Hill Notation):
C18H21NO3 · C4H6O6 · 2.5H2O
CAS Number:
Molecular Weight:
494.49
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing