
Hydrochloric acid

Synonym(s):
Hydrochloric acid, Hydrogen chloride solution, Hydrochloric acid solution, Hydrochloric acid standard solution
Linear Formula:
HCl
CAS Number:
Molecular Weight:
36.46
Products
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique
Showing 1-20 of 45
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3