Skip to Content
Merck
Homatropine hydrobromide European Pharmacopoeia (EP) Reference Standard

Homatropine hydrobromide

Homatropine hydrobromide European Pharmacopoeia (EP) Reference Standard
Synonym(s):
DL-Homatropine hydrobromide, DL-endo-α-Hydroxybenzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester hydrobromide, Tropine mandelate hydrobromide
Empirical Formula (Hill Notation):
C16H21NO3 · HBr
CAS Number:
Molecular Weight:
356.25

Products

Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade