
Heptakis(2,6-di-O-methyl)-β-cyclodextrin

Synonym(s):
2,6-Di-O-methyl-β-cyclodextrin, Dimethyl β-cyclodextrin
Empirical Formula (Hill Notation):
C56H98O35
CAS Number:
Molecular Weight:
1331.36
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing