Skip to Content
Merck
HATU 97%

HATU

HATU 97%
Synonym(s):
1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate, N-[(Dimethylamino)-1H-1,2,3-triazolo-[4,5-b]pyridin-1-ylmethylene]-N-methylmethanaminium hexafluorophosphate N-oxide, HATU
Empirical Formula (Hill Notation):
C10H15N6O · F6P
CAS Number:
Molecular Weight:
380.23

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing