Skip to Content
Merck
Halosulfuron-methyl PESTANAL®, analytical standard

Halosulfuron-methyl

Halosulfuron-methyl PESTANAL®, analytical standard
Synonym(s):
Methyl 3-chloro-5-(4,6-dimethoxy-2-pyrimidinylcarbamoylsulfamoyl)-1-methylpyrazole-4-carboxylate
Empirical Formula (Hill Notation):
C13H15ClN6O7S
CAS Number:
Molecular Weight:
434.81

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique