![Glyceryl tri(palmitate-1-13C) endotoxin tested, 99 atom % 13C, 98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/739/968/8c2f76a2-d242-48a6-b959-afcb1b2c412e/640/8c2f76a2-d242-48a6-b959-afcb1b2c412e.png)
Glyceryl tri(palmitate-1-13C)
![Glyceryl tri(palmitate-1-13C) endotoxin tested, 99 atom % 13C, 98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/739/968/8c2f76a2-d242-48a6-b959-afcb1b2c412e/640/8c2f76a2-d242-48a6-b959-afcb1b2c412e.png)
Synonym(s):
1,2,3-propanetriol tris(hexadecanoate-1-13C), Glyceryl tri(hexadecanoate-1-13C), Tripalmitin-carboxyls-13C3
Linear Formula:
[CH3(CH2)1413CO2CH2]2CHO213C(CH2)14CH3
CAS Number:
Molecular Weight:
810.30
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing