
Flubendiamide

Synonym(s):
N2-[1,1-Dimethyl-2-(methylsulfonyl)ethyl]-3-iodo-N1-{2-methyl-4-[1,2,2,2-tetrafluoro-1-(trifluoromethyl)ethyl]phenyl}-1,2-benzenedicarboxamide
Empirical Formula (Hill Notation):
C23H22F7IN2O4S
CAS Number:
Molecular Weight:
682.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2