
Ethoxyquin

Synonym(s):
1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, Ethoxyquin
Empirical Formula (Hill Notation):
C14H19NO
CAS Number:
Molecular Weight:
217.31
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3