
Escitalopram Related Compound A

Synonym(s):
(S)-4-[4-(Dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl]-3-hydroxymethylbenzonitrile hemi(+)-di-toluoyltartrate
Linear Formula:
C20H23FN2O2 · 0.5C20H18O8
CAS Number:
Molecular Weight:
535.58
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2