Skip to Content
BM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Centrifuge 5425
Synonym(s):
Eppendorf
®
Centrifuge 5425
Products
Product Category
centrifuges (5)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
EP5405000441
Eppendorf
®
Centrifuge 5425,
knob control, with Rotor FA-24x2 including aerosol-tight QuickLock lid, AC/DC input 120 V AC, US 3-pin plug
Expand
View Pricing
EP5405000107
Eppendorf
®
Centrifuge 5425,
refrigerated: no, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug
Expand
View Pricing
EP5405000646
Eppendorf
®
Centrifuge 5425,
knob control, without rotor, AC/DC input 120 V
Expand
View Pricing
EP5405000042
Eppendorf
®
Centrifuge 5425,
keypad control, with Rotor FA-24x2 including aerosol-tight QuickLock lid, AC/DC input 120 V
Expand
View Pricing
EP5405000247
Eppendorf
®
Centrifuge 5425,
keypad control, without rotor, AC/DC input 120 V
Expand
View Pricing