
Diethyl aminomalonate hydrochloride

Synonym(s):
Aminomalonic acid diethyl ester hydrochloride
Linear Formula:
NH2CH2(COOC2H5)2 · HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing