
Dabigatran impurity A CRS

Synonym(s):
(Ethyl 3-(2-{[(4-{[(hexyloxy)carbonyl]carbamoyl}phenyl)amino]methyl}-1-methyl-N-(pyridin-2-yl)-1H-benzimidazole-5-carboxamido)propanoate)
Empirical Formula (Hill Notation):
C34H40N6O6
CAS Number:
Molecular Weight:
628.72
Products
Formula Weight
Manufacturer
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing